5-Chloro-2-nitrophenol

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40511G5EA 0 CHF
ACTIAS40511G5
5-Chloro-2-nitrophenol
5-Chloro-2-nitrophenol
Formula: C₆H₄ClNO₃
MW: 173.56 g/mol
MDL Number: MFCD09260849
CAS Number: 611-07-4
EINECS: 210-249-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR