4-Bromo-o-cresol

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 4-Bromo-2-methylphenol1-Bromo-4-hydroxy-3-methylbenzene

ACTIAS40399G100EA 0 CHF
ACTIAS40399G100
4-Bromo-o-cresol
4-Bromo-o-cresol
Formula: C₇H₇BrO
MW: 187.04 g/mol
MDL Number: MFCD00055435
CAS Number: 2362-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR