2-Bromo-5-chloroanisole

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1-Bromo-4-chloro-2-methoxybenzene

ACTIAS40182G10EA 0 CHF
ACTIAS40182G10
2-Bromo-5-chloroanisole
2-Bromo-5-chloroanisole
Formula: C₇H₆BrClO
MW: 221.48 g/mol
MDL Number: MFCD03790889
CAS Number: 174913-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR