4-Bromobenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS22368G100EA 50 CHF
ACTIAS22368G25 ACTIAS22368G100
4-Bromobenzyl alcohol
4-Bromobenzyl alcohol
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 77…78 °C
MDL Number: MFCD00004650
CAS Number: 873-75-6
EINECS: 212-851-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR