3-Bromo-5-fluorotoluene

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS32308G5EA 28 CHF
ACTIAS32308G5 ACTIAS32308G10
3-Bromo-5-fluorotoluene
3-Bromo-5-fluorotoluene
Formula: C₇H₆BrF
MW: 189.03 g/mol
MDL Number: MFCD01861195
CAS Number: 202865-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR