2-Brom-4-chlortoluol

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

Synonyms: 1-Brom-3-chlor-6-methylbenzol

ACTIAS24941G100EA 122 CHF
ACTIAS24941G100
2-Brom-4-chlortoluol
2-Brom-4-chlortoluol
Formel: C₇H₆BrCl
MW: 205,48 g/mol
MDL Number: MFCD00060650
CAS-Nummer: 27139-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR