4-Amino-3-nitrophenol

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Hydroxy-2-nitroaniline

ACTIAS21438G100EA 0 CHF
ACTIAS21438G100
4-Amino-3-nitrophenol
4-Amino-3-nitrophenol
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 149…152 °C
MDL Number: MFCD00066310
CAS Number: 610-81-1
EINECS: 210-236-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR