4-Fluoro-o-toluidine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Amino-5-fluorotoluene4-Fluoro-2-methylaniline

ACTIAS31489G100EA 83 CHF
ACTIAS31489G100
4-Fluoro-o-toluidine
4-Fluoro-o-toluidine
Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: 14 °C
Density: 1.1263 g/cm³ (18 °C)
MDL Number: MFCD00007832
CAS Number: 452-71-1
EINECS: 207-208-2
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR