2-Amino-5-fluorophenol

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31682G5EA 0 CHF
ACTIAS31682G5 ACTIAS31682G1
2-Amino-5-fluorophenol
2-Amino-5-fluorophenol
Formula: C₆H₆FNO
MDL Number: MFCD00671759
CAS Number: 53981-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR