4-Chloro-2-(trifluoromethyl)aniline

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Amino-5-chloro-α,α,α-trifluorotoluene2-Amino-5-chlorobenzotrifluoride2-Amino-5-chloro-α,α,α-trifluortoluene4-Chloro-α,α,α-trifluoro-o-toluidine

ACTIAS40157G100EA 37.35 CHF
ACTIAS40157G100
4-Chloro-2-(trifluoromethyl)aniline
4-Chloro-2-(trifluoromethyl)aniline
Formula: C₇H₅ClF₃N
MW: 195.57 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007841
CAS Number: 445-03-4
EINECS: 207-151-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR