5-Bromo-2-fluoroaniline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS32338G100EA 123.91 CHF
ACTIAS32338G100
5-Bromo-2-fluoroaniline
5-Bromo-2-fluoroaniline
Formula: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD03094181
CAS Number: 2924-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR