3-Bromo-4-fluoroaniline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40320G100EA 0 CHF
ACTIAS40320G100
3-Bromo-4-fluoroaniline
3-Bromo-4-fluoroaniline
Formula: C₆H₅BrFN
MDL Number: MFCD03788559
CAS Number: 656-64-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR