3-(Trifluoromethoxy)aniline

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 3-Trifluoromethoxyaniline

ACTIAS20318G25EA 0 CHF
ACTIAS20318G25 ACTIAS20318G100
3-(Trifluoromethoxy)aniline
3-(Trifluoromethoxy)aniline
Formula: C₇H₆F₃NO
MW: 177.13 g/mol
Boiling Pt: 72 °C (1013 hPa)
Density: 1.3254 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041511
CAS Number: 1535-73-5
EINECS: 216-256-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR