3-(Difluoromethoxy)aniline

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS32320G5EA 0 CHF
ACTIAS32320G5 ACTIAS32320G10
3-(Difluoromethoxy)aniline
3-(Difluoromethoxy)aniline
Formula: C₇H₇F₂NO
MW: 159.14 g/mol
MDL Number: MFCD00236221
CAS Number: 22236-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR