6-Nitro-o-toluidine

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: 6-Nitro-2-methylaniline2-Methyl-6-nitroaniline2-Amino-3-nitrotoluene

ACTIAS21114G25EA 0 CHF
ACTIAS21114G25 ACTIAS21114G100
6-Nitro-o-toluidine
6-Nitro-o-toluidine
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 120…124 °C (1 mmHg)
Melting Pt: 92…94 °C
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007744
CAS Number: 570-24-1
EINECS: 209-329-6
UN: 2660
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR