2-Fluoro-6-nitroaniline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS32292G1EA 0 CHF
ACTIAS32292G1 ACTIAS32292G5
2-Fluoro-6-nitroaniline
2-Fluoro-6-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
MDL Number: MFCD04112583
CAS Number: 17809-36-8
EINECS: 241-779-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR