α-Chloro-2,4,5-trifluorotoulene

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: 2,4,5-Trifluorobenzyl chloride1-(Chloromethyl)-2,4,5-trifluorobenzene

ACTIAS32811G5EA 0 CHF
ACTIAS32811G5 ACTIAS32811G10
α-Chloro-2,4,5-trifluorotoulene
α-Chloro-2,4,5-trifluorotoulene
Formula: C₇H₄ClF₃
MW: 180.56 g/mol
MDL Number: MFCD00236319
CAS Number: 243139-71-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR