4-Methylbenzylamine

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: 4-(Aminomethyl)toluene

ACTIAS32943G10EA 0 CHF
ACTIAS32943G10
4-Methylbenzylamine
4-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: 20…21 °C
Density: 0.952 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008123
CAS Number: 104-84-7
EINECS: 203-243-2
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR