4-Chlorobenzylamine

Warning

Synonyms: (4-Chlorophenyl)methanamine

ACTIAS21175G100EA 0 CHF
ACTIAS21175G25 ACTIAS21175G100
4-Chlorobenzylamine
4-Chlorobenzylamine
Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 215 °C (1013 hPa)
Density: 1.16 g/cm³ (20 °C)
MDL Number: MFCD00008121
CAS Number: 104-86-9
EINECS: 203-245-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR