3-Methoxybenzylamine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS33228G5EA 0 CHF
ACTIAS33228G5 ACTIAS33228G10
3-Methoxybenzylamine
3-Methoxybenzylamine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 238 °C (1013 hPa)
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00008115
CAS Number: 5071-96-5
EINECS: 225-779-6
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR