3,5-Dichlorobenzylamine
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Danger
|
ACTIAS32838G1EA
0
CHF
ACTIAS32838G1
ACTIAS32838G10
ACTIAS32838G5
3,5-Dichlorobenzylamine
3,5-Dichlorobenzylamine
Formula:
C₇H₇Cl₂N MW: 176.04 g/mol Boiling Pt: 254 °C (6 mmHg) Density: 1.32 g/cm³ (20 °C) |
MDL Number:
MFCD00052681 CAS Number: 39989-43-0 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...