Suberic acid

Warning

Synonyms: Octanedioic acid1,6-Hexanedicarboxylic acid1,8-Octanedioic acid

ACTIAS21351G100EA 0 CHF
ACTIAS21351G100
Suberic acid
Suberic acid
Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 361 °C (1013 hPa)
Melting Pt: 139…142 °C
Density: 1,354 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004428
CAS Number: 505-48-6
EINECS: 208-010-9
Merck Index: 12,09031

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR