2',5'-Dichloroacetophenone

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS20677G100EA 0 CHF
ACTIAS20677G100
2',5'-Dichloroacetophenone
2',5'-Dichloroacetophenone
Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 56…58 °C (0.09 mmHg)
Melting Pt: 14 °C
Density: 1.352 g/cm³ (17.5 °C)
MDL Number: MFCD00000607
CAS Number: 2476-37-1
EINECS: 219-605-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR