2,5-Dichloro-α,α,α-trifluorotoluene

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: 1,4-Dichloro-2-(trifluoromethyl)benzene2,5-Dichlorobenzotrifluoride

ACTIAS24779G25EA 0 CHF
ACTIAS24779G25 ACTIAS24779G100
2,5-Dichloro-α,α,α-trifluorotoluene
2,5-Dichloro-α,α,α-trifluorotoluene
Formula: C₇H₃Cl₂F₃
MW: 215 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 11 °C
Density: 1.5009 g/cm³ (20 °C)
MDL Number: MFCD00000608
CAS Number: 320-50-3
EINECS: 206-276-0
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR