α,α,α,2,3-Pentafluortoluol
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Gefahr
|
ACTIAS24188G25EA
0
CHF
ACTIAS24188G25
ACTIAS24188G100
α,α,α,2,3-Pentafluortoluol
α,α,α,2,3-Pentafluortoluol
Formel:
C₇H₃F₅ MW: 182,09 g/mol |
MDL Number:
MFCD01631429 CAS-Nummer: 64248-59-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...