Tetrafluorophthalonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1,2-Dicyano-3,4,5,6-tetrafluorobenzene

ACTIAS24724G25EA 0 CHF
ACTIAS24724G100 ACTIAS24724G25
Tetrafluorophthalonitrile
Tetrafluorophthalonitrile
Formula: C₈F₄N₂
MW: 200.1 g/mol
MDL Number: MFCD00001774
CAS Number: 1835-65-0
EINECS: 217-400-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR