(4-Bromphenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS20130G100EA 59 CHF
ACTIAS20130G100
(4-Bromphenyl)essigsäure
(4-Bromphenyl)essigsäure
Formel: (C₆H₄Br)CH₂COOH
MW: 215,05 g/mol
Siedepunkt: 326 °C (1013 hPa)
Melting Pt: 94…98 °C
MDL Number: MFCD00004342
CAS-Nummer: 1878-68-8
EINECS: 217-523-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR