(2-Fluorophenyl)acetic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-(2-Fluorophenyl)acetic acid

ACTIAS20666G100EA 0 CHF
ACTIAS20666G100
(2-Fluorophenyl)acetic acid
(2-Fluorophenyl)acetic acid
Formula: (C₆H₄F)CH₂COOH
MW: 154.14 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 64 °C
MDL Number: MFCD00004315
CAS Number: 451-82-1
EINECS: 207-196-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR