(2,6-Dichlorophenyl)acetic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 2,6-Dichlorophenylacetic acid

ACTIAS20490G100EA 164 CHF
ACTIAS20490G100
(2,6-Dichlorophenyl)acetic acid
(2,6-Dichlorophenyl)acetic acid
Formula: (C₆H₃Cl₂)CH₂COOH
MW: 205.04 g/mol
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 160…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00004320
CAS Number: 6575-24-2
EINECS: 229-504-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR