3-Methylisoquinoline

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS40361G5EA 0 CHF
ACTIAS40361G5
3-Methylisoquinoline
3-Methylisoquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 66…67 °C
MDL Number: MFCD00024139
CAS Number: 1125-80-0
EINECS: 214-412-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR