2-Methylindole

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS40255G100EA 0 CHF
ACTIAS40255G100
2-Methylindole
2-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 56…59 °C
Density: 1,07 g/cm³ (20 °C)
Flash Pt: 141 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005616
CAS Number: 95-20-5
EINECS: 202-398-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR