(S)-(+)-2-Methylpiperazine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: (S)-2-Methylpiperazine

ACTIAS32954G5EA 0 CHF
ACTIAS32954G5 ACTIAS32954G25
(S)-(+)-2-Methylpiperazine
(S)-(+)-2-Methylpiperazine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 155…156 °C (1013 hPa)
MDL Number: MFCD00171452
CAS Number: 74879-18-8
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR