1,2,4-Trihydroxybenzene

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1,2,4-BenzenetriolBenzene-1,2,4-triol

ACTIAS21244G25EA 83 CHF
ACTIAS21244G25 ACTIAS21244G100
1,2,4-Trihydroxybenzene
1,2,4-Trihydroxybenzene
Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 362 °C (1013 hPa)
Melting Pt: 140 °C
Density: 1.45 g/cm³ (20 °C)
MDL Number: MFCD00002198
CAS Number: 533-73-3
EINECS: 208-575-1
Merck Index: 13,01073

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR