(S)-(+)-1,2-Propanediol

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: (S)-(+)-1,2-Dihydroxy propane(S)-(+)-1,2-Propylene glykol(S)-Propane-1,2-diol

ACTIAS21000G25EA 0 CHF
ACTIAS21000G25 ACTIAS21000G100
(S)-(+)-1,2-Propanediol
(S)-(+)-1,2-Propanediol
Formula: C₃H₈O₂
MW: 76.1 g/mol
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004539
CAS Number: 4254-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR