5-Methoxyindole

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS50146G25EA 0 CHF
ACTIAS50146G25
5-Methoxyindole
5-Methoxyindole
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 312 °C (1013 hPa)
Melting Pt: 55…56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005674
CAS Number: 1006-94-6
EINECS: 213-745-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR