D(-)-Leucinol

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: D-Leucinol(R)-(-)-2-Amino-4-methylpentan-1-ol(R)-(-)-Leucinol

ACTIAS32747G5EA 0 CHF
ACTIAS32747G5 ACTIAS32747G1
D(-)-Leucinol
D(-)-Leucinol
Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 199 °C (1013 hPa)
Density: 0.897 g/cm³ (20 °C)
CAS Number: 53448-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR