3,4,5-Trimethylaniline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS37124G1EA 0 CHF
ACTIAS37124G1 ACTIAS37124MG250
3,4,5-Trimethylaniline
3,4,5-Trimethylaniline
Formula: C₉H₁₃N
CAS Number: 1639-31-2
EINECS: 216-679-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR