2-(Trifluoromethoxy)aniline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS21174G100EA 0 CHF
ACTIAS21174G100
2-(Trifluoromethoxy)aniline
2-(Trifluoromethoxy)aniline
Formula: C₇H₆F₃NO
MW: 177.13 g/mol
Boiling Pt: 64.5 °C (21 mmHg)
Density: 1.301 g/cm³ (20 °C)
MDL Number: MFCD00035959
CAS Number: 1535-75-7
EINECS: 216-257-9
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR