2-Amino-3-fluorpyridin
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS30503G1EA
0
CHF
ACTIAS30503G1
ACTIAS30503G5
2-Amino-3-fluorpyridin
2-Amino-3-fluorpyridin
Formel:
C₅H₅FN₂ MW: 112,11 g/mol |
MDL Number:
MFCD04114135 CAS-Nummer: 21717-95-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...