6-Fluoro-1-indanone

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS31490G5EA 0 CHF
ACTIAS31490G5 ACTIAS31490G1
6-Fluoro-1-indanone
6-Fluoro-1-indanone
Formula: C₉H₇FO
MW: 150.15 g/mol
MDL Number: MFCD01318147
CAS Number: 1481-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR