3,3-Dimethylglutaric acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3,3-Dimethylpentanedioic acid3,3-Dimethyl-1,5-pentanedioic acid

ACTIAS40274G100EA 0 CHF
ACTIAS40274G100
3,3-Dimethylglutaric acid
3,3-Dimethylglutaric acid
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 98…101 °C
Density: 1.4278 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002716
CAS Number: 4839-46-7
EINECS: 225-425-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR