D-(+)-Tryptophan

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: H-D-Trp-OH

ACTIAS33044G100EA 0 CHF
ACTIAS33044G100
D-(+)-Tryptophan
D(+)-Tryptophan
Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 280…283 °C
MDL Number: MFCD00005647
CAS Number: 153-94-6
EINECS: 205-819-9
Merck Index: 14,09797

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR