Cesium acetate

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS21473G100EA 0 CHF
ACTIAS21473G100
Cesium acetate
Cesium acetate
Formula: H₃CCOOCs
MW: 191.95 g/mol
Melting Pt: 194 °C
MDL Number: MFCD00013056
CAS Number: 3396-11-0
EINECS: 222-248-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR