3-Acetylpyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Methyl 3-pyridyl ketone1-(Pyridin-3-yl)ethanone

ACTIAS20114G100EA 0 CHF
ACTIAS20114G100
3-Acetylpyridine
3-Acetylpyridine
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 220 °C (1013 hPa)
Density: 1.1065 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006396
CAS Number: 350-03-8
EINECS: 206-496-7
UN: 2810
ADR: 6.1,III
Merck Index: 12,06195

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR