Indole-4-carbonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 1H-Indole-4-carbonitrile4-Cyano-1H-indole4-Cyanoindole

ACTIAS7190G100EA 0 CHF
ACTIAS7190G5 ACTIAS7190G100 ACTIAS7190G25
Indole-4-carbonitrile
Indole-4-carbonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
MDL Number: MFCD00152045
CAS Number: 16136-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR