2-Amino-6-chloro-3-nitropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 6-Chloro-3-nitro-2-pyridylamine6-Chloro-3-nitropyridin-2-amine

ACTIAS7081G5EA 0 CHF
ACTIAS7081G25 ACTIAS7081G5 ACTIAS7081G1
2-Amino-6-chloro-3-nitropyridine
2-Amino-6-chloro-3-nitropyridine
Formula: C₅H₄ClN₃O₂
MW: 173.56 g/mol
Melting Pt: 195…196 °C
MDL Number: MFCD00044350
CAS Number: 27048-04-0
EINECS: 248-188-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR