2-Chloro-6-nitroaniline

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

ACTIAS31355G1EA 0 CHF
ACTIAS31355G10 ACTIAS31355G1
2-Chloro-6-nitroaniline
2-Chloro-6-nitroaniline
Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Storage Temperature: Ambient
CAS Number: 769-11-9
EINECS: 212-204-9
UN: 2237
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR