3-Chlor-4-hydroxybenzoesäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
ACTIAS20369G25EA
0
CHF
ACTIAS20369G50
ACTIAS20369G250
ACTIAS20369G25
3-Chlor-4-hydroxybenzoesäure
3-Chloro-4-hydroxybenzoic acid
Formel:
C₇H₅ClO₃ MW: 172,57 g/mol Siedepunkt: 331 °C (1013 hPa) |
MDL Number:
MFCD00002549 CAS-Nummer: 3964-58-7 EINECS: 223-574-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...