6-Chloro-m-toluic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS40214G5EA 0 CHF
ACTIAS40214G5 ACTIAS40214G25
6-Chloro-m-toluic acid
6-Chloro-m-toluic acid
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
MDL Number: MFCD00045798
CAS Number: 6342-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR