2-Bromo-5-hydroxypyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 2-Bromo-5-pyridinol6-Bromopyridin-3-ol

ACTIAS4051G5EA 0 CHF
ACTIAS4051G5
2-Bromo-5-hydroxypyridine
2-Bromo-5-hydroxypyridine
Formula: Câ‚…Hâ‚„BrNO
MW: 174 g/mol
MDL Number: MFCD04114184
CAS Number: 55717-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR